Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methyl-2-nitrobenzenesulphonamide
N-Methyl-2-nitrobenzenesulphonamide
N-Methyl-2-nitrobenzenesulphonamide
N-Methyl-2-nitrobenzenesulphonamide
APOSOR12494-1G
:
Apollo Scientific
:
N-Methyl-2-nitrobenzenesulphonamide
APOSOR12494-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
216.2100 g/mol
:
109…111 °C
:
GHS07
:
Ambient
:
23530-40-7
:
C₇H₈N₂O₄S
:
MFCD01115621