Order Entry
Export
ContactUsLinkComponent
3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde
3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde
Catalog # APOSOR12476-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-Phenyl-5,6-dihydroimidazo[2,1-b][1,3]thiazole-2-carbaldehyde
Catalog # APOSOR12476-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :230.29 g/mol
Melting Pt:138…139 °C
Storage Temperature:Ambient
CAS Number:952182-40-0
Formula:C₁₂H₁₀N₂OS
MDL Number:MFCD09817450