
2,4-Dibromo-1-methylimidazole 98%
Catalog # APOSOR1228-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:239.9 g/mol
Melting Pt:74…79 °C
:GHS07
Storage Temperature:Ambient
CAS Number:53857-60-6
Formula:C₄H₄Br₂N₂
MDL Number:MFCD06659906