Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-1-methylimidazole
4-Bromo-1-methylimidazole 98%
4-Bromo-1-methylimidazole 98%
:
Apollo Scientific
4-Bromo-1-methylimidazole 98%
:
Apollo Scientific
:
25676-75-9
:
:
C₄H₅BrN₂
:
MFCD01320501
:
161 g/mol
:
91…93 °C/0.1mmHg
:
1.614 g/cm³ (25 °C)
:
110 °C
:
GHS07
:
Ambient