2-Chloro-N-(2,3-dimethylphenyl)acetamide
Catalog # APOSOR12177-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:197.66 g/mol
Melting Pt:135…136 °C
Storage Temperature:Ambient
CAS Number:2564-07-0
Formula:C₁₀H₁₂ClNO
MDL Number:MFCD00018895