Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Bromo-4,5-dichloro-2-nitrobenzene
1-Bromo-4,5-dichloro-2-nitrobenzene
1-Bromo-4,5-dichloro-2-nitrobenzene
Irritant
1-Bromo-4,5-dichloro-2-nitrobenzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1-Bromo-4,5-dichloro-2-nitrobenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
93361-94-5
:
Formula:
C₆H₂BrCl₂NO₂
MDL Number:
MFCD07780670
:
270.9000 g/mol
Melting Pt:
60…64 °C
:
GHS07
Storage Temperature:
Ambient