
Methyl imidazo[1,2-a]pyridine-6-carboxylate hydrochloride
Catalog # APOSOR11947-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:957120-91-1
Formula:C₉H₉ClN₂O₂
MDL Number:MFCD09801003