Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloro-m-toluidine
2,6-Dichloro-m-toluidine 90+%
2,6-Dichloro-m-toluidine 90+%
:
Apollo Scientific
2,6-Dichloro-m-toluidine 90+%
:
Apollo Scientific
:
64063-37-2
:
:
C₇H₇Cl₂N
:
MFCD00007676
:
176.0432 g/mol
:
37…40 °C
:
GHS07
:
Refrigerator