Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromobenzene-1,2-diamine
3-Bromobenzene-1,2-diamine, Technical Grade
3-Bromobenzene-1,2-diamine, Technical Grade
Irritant
3-Bromobenzene-1,2-diamine, Technical Grade
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Bromobenzene-1,2-diamine, Technical Grade
Supplier:
Apollo Scientific
About this item
CAS Number:
1575-36-6
:
Formula:
C6H7BrN2
MDL Number:
MFCD09842434
:
187.0372 g/mol
Melting Pt:
53…55 °C
:
GHS07
Storage Temperature:
Ambient