2,4-Dihydroxybenzaldehyde
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:95-01-2
- Formula:C₇H₆O₃
- MDL Number:MFCD00011686
- Molecular Weight:138.12 g/mol
- Boiling Pt:220 °C
- Melting Pt:134…138 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:95-01-2
- Formula:C₇H₆O₃
- MDL Number:MFCD00011686
- Molecular Weight:138.12 g/mol
- Boiling Pt:220 °C
- Melting Pt:134…138 °C
- Storage Temperature:Ambient



