Order Entry
Export
ContactUsLinkComponent
Imidazo[1,2-a]pyridin-8-ol hydrochloride 97%
Irritant
Imidazo[1,2-a]pyridin-8-ol hydrochloride 97%
Catalog # APOSOR11533-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Imidazo[1,2-a]pyridin-8-ol hydrochloride 97%
Catalog # APOSOR11533-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:100592-11-8
Formula:C₇H₇ClN₂O
MDL Number:MFCD09800942