
3-(3,5-Dimethoxyphenyl)propionic acid
Catalog # APOSOR11325-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:210.23 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:717-94-2
Formula:C₁₁H₁₄O₄
MDL Number:MFCD03425685