Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dichloropurine

2,6-Dichloropurine

Irritant
New Product
Supplier:   Apollo Scientific
2,6-Dichloropurine
Supplier:  Apollo Scientific
New Product
Certificates
2,6-Dichloropurine

4 Options Available Below

Product Details & Documents
Synonyms:  2,6-Dichloro-9H-purine
  • CAS Number:5451-40-1
  • Formula:C₅H₂Cl₂N₄
  • MDL Number:MFCD00077725
  • Molecular Weight:189 g/mol
  • Melting Pt:184…186 °C
  • Storage Temperature:Ambient
  • EINECS:226-681-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 31,90 €
    31,90 €
    1 * 25 g
       31,90 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 126,00 €
    126,00 €
    1 * 100 g
       126,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 624,00 €
    624,00 €
    1 * 500 g
       624,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 3 110,00 €
    3 110,00 €
    1 * 2,5 kg
       3 110,00 €

    Specifications:
    New Product
    New Product