2-Hydroxypyridine
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
- CAS Number:142-08-5
- Formula:C₅H₅NO
- MDL Number:MFCD00006268
- Molecular Weight:95.10 g/mol
- Boiling Pt:280…281 °C
- Melting Pt:105…109 °C
- Storage Temperature:Ambient
- EINECS:205-520-3
- UN:2811
- ADR:6.1,III
6 Options Available Below
Product Details & Documents
Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
- CAS Number:142-08-5
- Formula:C₅H₅NO
- MDL Number:MFCD00006268
- Molecular Weight:95.10 g/mol
- Boiling Pt:280…281 °C
- Melting Pt:105…109 °C
- Storage Temperature:Ambient
- EINECS:205-520-3
- UN:2811
- ADR:6.1,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



