N-(5-Bromo-4-methyl-1,3-thiazol-2-yl)acetamide
Catalog # APOSOR111616-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.11 g/mol
Storage Temperature:Ambient
CAS Number:21478-95-5
Formula:C₆H₇BrN₂OS
MDL Number:MFCD01924770