Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichloro-6-methylquinoxaline
2,3-Dichloro-6-methylquinoxaline
2,3-Dichloro-6-methylquinoxaline
Irritant
2,3-Dichloro-6-methylquinoxaline
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Dichloro-6-methylquinoxaline
Supplier:
Apollo Scientific
About this item
CAS Number:
39267-05-5
:
Formula:
C₉H₆Cl₂N₂
MDL Number:
MFCD00270819
:
213.06 g/mol
:
GHS07
Storage Temperature:
Ambient