1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanol
Catalog # APOSOR111548-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:268.4 g/mol
Storage Temperature:Ambient
CAS Number:117553-23-8
Formula:C₁₉H₂₄O
MDL Number:MFCD18089273