
3-Bromoimidazo[1,2-a]pyridine-6-carboxylic acid
Catalog # APOSOR111401-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:241.05 g/mol (anhy)
:GHS07
Storage Temperature:Ambient
CAS Number:886362-00-1
Formula:C₈H₅BrN₂O₂
MDL Number:MFCD07021511