
4-(1,3-Dioxolan-2-yl)aniline
Catalog # APOSOR111317-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:165.19 g/mol
Boiling Pt:150 °C/1mmHg
Melting Pt:74…79 °C
:GHS07
Storage Temperature:Ambient
CAS Number:19073-14-4
Formula:C₉H₁₁NO₂
MDL Number:MFCD00604220