Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone
Supplier:
Apollo Scientific
Certificates
Expand image
1-(4'-Pentyl-1,1'-biphenyl-4-yl)ethanone
Supplier:
Apollo Scientific
About this item
CAS Number:
59662-38-3
:
Formula:
C₁₉H₂₂O
MDL Number:
MFCD00428365
:
266.39 g/mol
Storage Temperature:
Ambient