
1,3-Bis(4-chlorophenyl)urea
Catalog # APOSOR11102-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:281.14 g/mol
Melting Pt:302…305 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1219-99-4
Formula:C₁₃H₁₀Cl₂N₂O
MDL Number:MFCD00018541