Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
Irritant
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-(2-Methyl-1,3-thiazol-4-yl)benzonitrile
Supplier:
Apollo Scientific
About this item
CAS Number:
127406-79-5
:
Formula:
C₁₁H₈N₂S
MDL Number:
MFCD01765038
:
200.26 g/mol
:
GHS07
Storage Temperature:
Ambient