
3-Bromoimidazo[1,2-a]pyridin-8-amine
Catalog # APOSOR110820-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:212.05 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1232431-81-0
Formula:C₇H₆BrN₃
MDL Number:MFCD17012030