Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(6-Chloroimidazo[1,2-a]pyrid-2-yl)acetic acid
2-(6-Chloroimidazo[1,2-a]pyrid-2-yl)acetic acid
2-(6-Chloroimidazo[1,2-a]pyrid-2-yl)acetic acid
2-(6-Chloroimidazo[1,2-a]pyrid-2-yl)acetic acid
Supplier:
Apollo Scientific
Certificates
Expand image
2-(6-Chloroimidazo[1,2-a]pyrid-2-yl)acetic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
59128-13-1
:
Formula:
C₉H₇ClN₂O₂
MDL Number:
MFCD06739270
Storage Temperature:
Ambient