Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Phenyl-1,3-cyclohexanedione
5-Phenyl-1,3-cyclohexanedione 98%
5-Phenyl-1,3-cyclohexanedione 98%
:
Apollo Scientific
5-Phenyl-1,3-cyclohexanedione 98%
:
Apollo Scientific
:
493-72-1
:
:
C₁₂H₁₂O₂
:
MFCD00051846
:
188.23 g/mol
:
185…188 °C (dec.)
:
GHS07
:
Ambient