Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-1,3-benzoxazol-6-amine
2-Methyl-1,3-benzoxazol-6-amine
2-Methyl-1,3-benzoxazol-6-amine
:
Apollo Scientific
2-Methyl-1,3-benzoxazol-6-amine
:
Apollo Scientific
:
5676-60-8
:
:
C₁₁H₁₄N₂O
:
MFCD03791181
:
148.1619 g/mol
:
Ambient
:
2811
:
6.1,III
:
GHS06