Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-5-phenylthieno[2,3-d]pyrimidine
4-Chloro-5-phenylthieno[2,3-d]pyrimidine
4-Chloro-5-phenylthieno[2,3-d]pyrimidine
4-Chloro-5-phenylthieno[2,3-d]pyrimidine
Supplier:
Apollo Scientific
Certificates
Expand image
4-Chloro-5-phenylthieno[2,3-d]pyrimidine
Supplier:
Apollo Scientific
About this item
CAS Number:
182198-35-2
:
Formula:
C₁₂H₇ClN₂S
MDL Number:
MFCD00218029
:
246.72 g/mol
Storage Temperature:
Ambient