Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Imidazo[2,1-b][1,3]thiazol-6-ylaniline
4-Imidazo[2,1-b][1,3]thiazol-6-ylaniline
4-Imidazo[2,1-b][1,3]thiazol-6-ylaniline
Irritant
4-Imidazo[2,1-b][1,3]thiazol-6-ylaniline
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Imidazo[2,1-b][1,3]thiazol-6-ylaniline
Supplier:
Apollo Scientific
About this item
CAS Number:
183668-02-2
:
Formula:
C₁₁H₉N₃S
MDL Number:
MFCD01624149
:
215.28 g/mol
Melting Pt:
207…209 °C
:
GHS07
Storage Temperature:
Ambient