2-Methyl-4-phenoxyaniline
Catalog # APOSOR110537-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.25 g/mol
Storage Temperature:Ambient
CAS Number:13024-16-3
Formula:C₁₃H₁₃NO
MDL Number:MFCD08688317