Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Hydroxy-1-indanone
5-Hydroxy-1-indanone 97%
5-Hydroxy-1-indanone 97%
:
Apollo Scientific
5-Hydroxy-1-indanone 97%
:
Apollo Scientific
:
3470-49-3
:
:
C9H8O2
:
MFCD00857527
:
148.1600 g/mol
:
185…188 °C
:
GHS07
:
Ambient