
3-Bromo-8-nitroimidazo[1,2-a]pyridine
Catalog # APOSOR110437-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.04 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:52310-43-7
Formula:C₇H₄BrN₃O₂
MDL Number:MFCD17016106