Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-methyl-3-nitrobenzoate
Methyl-4-methyl-3-nitrobenzoate
Methyl-4-methyl-3-nitrobenzoate
APOSOR110363-25G
:
Apollo Scientific
:
Methyl-4-methyl-3-nitrobenzoate
APOSOR110363-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
195.18 g/mol
:
49…53 °C
:
>110 °C
:
230-886-6
:
GHS07
:
Ambient
:
7356-11-8
:
C₉H₉NO₄
:
MFCD00130054