(2-Phenyl-1,3-thiazol-5-yl)methanol
Catalog # APOSOR110315-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.25 g/mol
Storage Temperature:Ambient
CAS Number:859485-91-9
Formula:C₁₀H₉NOS
MDL Number:MFCD09694149