
6-Chloroimidazo[1,2-a]pyridine
Catalog # APOSOR110276-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:152.58 g/mol
Melting Pt:80…82 °C
:GHS07
Storage Temperature:Ambient
CAS Number:6188-25-6
Formula:C₇H₅ClN₂
MDL Number:MFCD05663807