Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-1-methyl-1H-pyrrole-2-carboxylate
Methyl-1-methyl-1H-pyrrole-2-carboxylate
Methyl-1-methyl-1H-pyrrole-2-carboxylate
Methyl-1-methyl-1H-pyrrole-2-carboxylate
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl-1-methyl-1H-pyrrole-2-carboxylate
Supplier:
Apollo Scientific
About this item
CAS Number:
37619-24-2
:
Formula:
C₇H₉NO₂
MDL Number:
MFCD00052747
:
139.16 g/mol
Storage Temperature:
Ambient