3-(Dimethylamino)-1-(1,3-thiazol-2-yl)-2-buten-1-one
Catalog # APOSOR110202-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.27 g/mol
Storage Temperature:Ambient
CAS Number:886361-88-2
Formula:C₉H₁₂N₂OS
MDL Number:MFCD08056641