
6-Bromo-2-methylimidazo[1,2-a]pyridine
Catalog # APOSOR110182-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:211.07 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
CAS Number:4044-99-9
Formula:C₈H₇BrN₂
MDL Number:MFCD09800594