Order Entry
Export
ContactUsLinkComponent
3-Bromo-7-chloroimidazo[1,2-a]pyridine
Irritant
3-Bromo-7-chloroimidazo[1,2-a]pyridine
Catalog # APOSOR110071-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Bromo-7-chloroimidazo[1,2-a]pyridine
Catalog # APOSOR110071-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :231.4800 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:342613-67-6
Formula:C₇H₄BrClN₂
MDL Number:MFCD11111834