Order Entry
Export
ContactUsLinkComponent
7-Chloroimidazo[1,2-a]pyridine
Irritant
7-Chloroimidazo[1,2-a]pyridine
Catalog # APOSOR110058-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
7-Chloroimidazo[1,2-a]pyridine
Catalog # APOSOR110058-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :152.5800 g/mol
Melting Pt:49…52 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:4532-25-6
Formula:C₇H₅ClN₂
MDL Number:MFCD08275126