p-Anisic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:100-09-4
:
Formula:C₈H₈O₃
MDL Number:MFCD00002542
:152.15 g/mol
Boiling Pt:275…280 °C
Melting Pt:182…185 °C
Density:1.385 g/cm³ (25 °C)
Flash Pt:185 °C
EINECS:202-818-5
:GHS07
Storage Temperature:Ambient