Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichlorobenzoic acid
2,3-Dichlorobenzoic acid
2,3-Dichlorobenzoic acid
Irritant
2,3-Dichlorobenzoic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,3-Dichlorobenzoic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
50-45-3
:
Formula:
C₇H₄Cl₂O₂
MDL Number:
MFCD00002413
Melting Pt:
168…170 °C
:
191.01 g/mol
EINECS:
200-039-5
:
GHS07
Storage Temperature:
Ambient