Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichlorobenzaldehyde
2,3-Dichlorobenzaldehyde 97%
2,3-Dichlorobenzaldehyde 97%
:
Apollo Scientific
2,3-Dichlorobenzaldehyde 97%
:
Apollo Scientific
:
6334-18-5
:
:
C₇H₄Cl₂O
:
MFCD00010127
:
175.01 g/mol
:
143…145 °C/30mmHg
:
62…64 °C
:
1.33 g/cm³ (25 °C)
:
135 °C
:
Ambient
:
228-711-3
:
3261
:
8,II
:
GHS05