Order Entry
Export
ContactUsLinkComponent
4,6-Dihydroxy-5-nitropyrimidine
4,6-Dihydroxy-5-nitropyrimidine
Catalog # APOSOR10855-25G
Supplier:  Apollo Scientific
CAS Number:  
4,6-Dihydroxy-5-nitropyrimidine
Catalog # APOSOR10855-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :157.09 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
CAS Number:2164-83-2
Formula:C₄H₃N₃O₄
MDL Number:MFCD00006110
Melting Pt:>300 °C