Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tri(2-furyl)phosphine
Tri(2-furyl)phosphine
Tri(2-furyl)phosphine
Tri(2-furyl)phosphine
Supplier:
Apollo Scientific
Certificates
Expand image
Tri(2-furyl)phosphine
Supplier:
Apollo Scientific
About this item
CAS Number:
5518-52-5
:
Formula:
C₁₂H₉O₃P
MDL Number:
MFCD00151857
:
232.17 g/mol
Boiling Pt:
136 °C/4mmHg
Melting Pt:
59…64 °C
:
GHS07
Storage Temperature:
Ambient