

4-Bromo-o-xylene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:583-71-1
:
Formula:C₈H₉Br
MDL Number:MFCD00000086
:185.06 g/mol
Boiling Pt:214…215 °C
Density:1.34 g/cm³ (25 °C)
Flash Pt:80 °C
EINECS:209-518-3
:GHS07
:GHS09
Storage Temperature:Ambient