Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dibromo-5-tert-butylbenzene
1,3-Dibromo-5-tert-butylbenzene
1,3-Dibromo-5-tert-butylbenzene
Irritant
1,3-Dibromo-5-tert-butylbenzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,3-Dibromo-5-tert-butylbenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
129316-09-2
:
Formula:
C₁₀H₁₂Br₂
MDL Number:
MFCD06657956
:
292.0100 g/mol
Melting Pt:
30…33 °C
:
GHS07
Storage Temperature:
Ambient