Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dimethylpyridin-4-ol
2,6-Dimethylpyridin-4-ol 98%
2,6-Dimethylpyridin-4-ol 98%
:
Apollo Scientific
2,6-Dimethylpyridin-4-ol 98%
:
Apollo Scientific
:
13603-44-6
:
:
C₇H₉NO
:
MFCD00956021
:
123.1525 g/mol
:
351 °C
:
230 °C
:
Ambient
:
237-089-2
:
GHS07