
Polyethylene glycol 1,000
Catalog # APOSOR1064-50G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:1000 g/mol
Melting Pt:61.7…66.6 °C
Density:1.21 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839