Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Methylthio)phenylboronic acid
3-(Methylthio)phenylboronic acid 97%
3-(Methylthio)phenylboronic acid 97%
Irritant
3-(Methylthio)phenylboronic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-(Methylthio)phenylboronic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
128312-11-8
:
Formula:
C₇H₉BO₂S
MDL Number:
MFCD01319105
Melting Pt:
158…160 °C
:
168.02 g/mol
:
GHS07
Storage Temperature:
Ambient