Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxy-2,6-dimethoxybenzaldehyde
4-Hydroxy-2,6-dimethoxybenzaldehyde 98%
4-Hydroxy-2,6-dimethoxybenzaldehyde 98%
Irritant
4-Hydroxy-2,6-dimethoxybenzaldehyde 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Hydroxy-2,6-dimethoxybenzaldehyde 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
22080-96-2
:
Formula:
C₉H₁₀O₄
MDL Number:
MFCD01407645
:
182.1733 g/mol
Melting Pt:
225 °C (dec.)
:
GHS07
Storage Temperature:
Ambient